Learn More
MAZ Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 21068-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
May function as a transcription factor with dual roles in transcription initiation and termination. Binds to two sites, ME1a1 and ME1a2, within the MYC promoter having greater affinity for the former. Also binds to multiple G/C-rich sites within the promoter of the Sp1 family of transcription factors.Specifica
| MAZ | |
| Polyclonal | |
| Unconjugated | |
| MAZ | |
| MAZ, MAZI, Pur 1, PUR1, SAF 1, SAF 2, ZF87, Zif87, ZNF801 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4150 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P56270 | |
| MAZ | |
| MAZ Fusion Protein Ag15200 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |