missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCCC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12117-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes the small subunit of 3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoA to form 3-methylglutaconyl-CoA. Mutations in this gene are associated with 3-Methylcrotonylglycinuria, an autosomal recessive disorder of leucine catabolism.Specifica
MCCC2 | |
Polyclonal | |
Unconjugated | |
MCCC2 | |
MCCase subunit beta, MCCB, MCCC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361884, 64087, 78038 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3ULD5, Q5XIT9, Q9HCC0 | |
Mccc2 | |
MCCC2 Fusion Protein Ag2767 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |