Learn More
MCTP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA562337
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: RRVSSNGIFD LQKTSLGGDA PEEPEKLCGS SDLNASMTSQ HFEEQSVPGE ASDGLSNLPS PFAYLLTIHL KEGRNLVVRD R Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 63%.
Might play a role in the development of cardiac outflow tract. [UniProt]Specifica
MCTP2 | |
Polyclonal | |
Unconjugated | |
MCTP2 | |
Gm489; MCTP2; multiple C2 and transmembrane domain containing 2; multiple C2 and transmembrane domain-containing protein 2; multiple C2 domains, transmembrane 2; multiple C2-domains with two transmembrane regions 2 1; RGD1562967 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DN12 | |
MCTP2 | |
Recombinant protein corresponding to Human MCTP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only