missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11049-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
11049-1-AP can recognize both MEK2 and MEK1.
MEK1 (Mitogen activated protein kinase kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.Specifica
MEK2 | |
Polyclonal | |
Unconjugated | |
MAP2K2 | |
ERK activator kinase 2, MAP kinase kinase 2, MAP2K2, MAPK/ERK kinase 2, MAPKK 2, MAPKK2, MEK 2, MEK2, MKK2, PRKMK2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26396, 5605, 58960 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36506, P36507, Q63932 | |
Map2k2 | |
MEK2 Fusion Protein Ag1481 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |