Learn More
Uvex™ Men's Jacket 7475-Night Blue
Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.
Marca:  Uvex™ 8828115
Dettagli aggiuntivi : Peso : 0.81500kg
Codice prodotto. 17245921
  • 238.00€ / Pezzo
Spedizione stimata: 26-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Jacket uvex suXXeed multifunction
  • Material: 49% PPAN-FR, 42% cotton, 5% Aramid, 3% Polyamid, 1% antistat. fibres, 300 gsm
  • Multifunctional, for hightest protection, soft and skin friendly, integrated stretch inserts, high rise Sleeve construction, reflective inserts
  • Standards: - EN ISO 11611 - EN ISO 11612 - EN 1149-5 - EN 13034 Type 6 - IEC 61482-1-2
  • Sizes: S - 6XL
  • Industrial wash, 60 degree, no bleaching, 2 points tumbler, 2 points ironing
Specifica
Specifica
4X-Large
Blue
No
Men's
Zipper
300 g/m²