Learn More
Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
Marca: Metro™ LT30XS3
Dettagli aggiuntivi : Peso : 21.77600kg
Descrizione
- Table features smooth surfaces that are easy to clean
- Mobile units can be easily moved to clean behind table area
- Available top materials include Type 304 stainless and TRESPA phenolic resin (black or gray)
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifica
29.75 in. | |
29.75 x 30 x 39.5 in. (74.9 x 76.2 x 100.3 cm) | |
100.3 cm | |
Backsplash with solid Metro™ MetroMax i™ shelf | |
273 kg | |
74.9 cm | |
Polymer |
Stationary Table | |
Ideal for lab | |
39.5 in. | |
30 in. | |
76.2 cm | |
600 lb. |