Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Black Phenolic Top with 3-Sided Frame
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Marca:  Metro™ LT36XUPB3
Dettagli aggiuntivi : Peso : 33.38400kg
Codice prodotto. 15977031
  • 1390.00€ / Pezzo
Spedizione stimata: 23-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
These worktables provide the ideal product platform, are modular by design and offer ultimate flexibility based on the application requirements.
  • Advanced polymer construction for corrosive lab environments
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifica
Specifica
35.75 in.
35.75 x 30 x 40.5 in. (90.8 x 76.2 x 101.6 cm)
101.6 cm
3 Sided frame
273 kg
90.8 cm
Polymer
Stationary Table
Ideal for lab
40.5 in.
30 in.
76.2 cm
600 lb.