Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Shelf Ledge, Stackable
Shelf ledges may be stacked on top of each other to create taller ledges, Ledges snap onto shelves and posts for secure fit with no tools
Marca: Metro™ L36N4C
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- 4 in. high (10cm)
- Choice of plated or stainless steel finish
Order Info
Posts and shelves sold separately.
Specifica
36 in. | |
91.4 cm | |
Metro™ Super Erecta™ wire shelving |
10 cm | |
4 in. | |
Chrome |