missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Rat, APC-Cy7, Clone: M5/114.15.2, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB70-APC7-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MHC Class II | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
H2-Ab1 and H2-Eb1 | |
Protein A/G Purified | |
RUO | |
14969 | |
IgG2b κ |
Flow Cytometry | |
M5/114.15.2 | |
APC-Cyanine7 | |
NP_034512 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |