Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with GN-6 Membrane
Ready-to-use, sterile and individually packaged MicroFunnel filter funnels reduce chances of cross contamination.
Marca: Cytiva (formerly Pall Lab) 4800
297.16 EUR valido fino al 2024-06-30
Usa il codice promo "21742" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.93800kg
Descrizione
- Easy squeeze removal of funnel to access membrane – no twisting means less RSI and membrane tearing
- Proven membrane performance
- For research use only
Specifica
100 mL | |
50/Pk. | |
Individually Wrapped |
0.45 μm | |
Sterile | |
GN-6 Metricel Mixed Cellulose Ester |