Learn More
Sartorius Stedim Biotech™ Microsart™ RESEARCH Bacteria
Codice prodotto. p-7084293 Sfoglia Tutto Sartorius Prodotti
Click to view available options
N. di reazioni:
Reagents for 100 PCR reactions
Reagents for 25 PCR reactions
Quantità:
25
100
Dimensione della confezione:
pezzi 100
pezzi 25
Codice prodotto. 15943030
Marca: Sartorius Stedim Biotech™ SMB951009
Sartorius Stedim Biotech™ Microsart™ RESEARCH Bacteria is used for fast and reliable real-time PCR-based detection of bacterial contamination in cell cultures or cell culture supernatants.
  • Rapidly detect contamination of more than 95 % of all known bacterial species within 2½ hours.
  • In a Real-time PCR highly specific TaqMan probes bind to a very conserved DNA-sequence that is common for all bacteria (gram-positive and gram negative).
  • This kit is designed for contamination control in academia or during drug development and in-process in biopharma or regenerative medicine.
TRUSTED_SUSTAINABILITY
Specifica
Metodo di rilevamento qPCR
Quantità 25
N. di reazioni Reagents for 25 PCR reactions
Volume 2μL