Learn More
Carl Zeiss™ Microscope Stage Micrometers
Finely divided stage micrometer for calibrating optical systems with eyepiece graticule patterns. Carl Zeiss™ Microscope Stage Micrometer includes 1mm and 1/100mm intervals.
Marca: Carl Zeiss™ 474027-0000-000
Dettagli aggiuntivi : Peso : 0.80000kg
Includes: 5 lines with 1mm intervals in y-direction, 100 lines with 1/100mm intervals in y-direction
Descrizione
- For calibrating optical systems with eyepiece graticule patterns
- 5 lines with 1mm intervals in y-direction
- 100 lines with 1/100mm intervals in y-direction
Specifica
Stage Micrometer | |
Reflected light microscope, eyepiece graticule | |
Stage micrometer, 5 lines with 1mm intervals in y-direction |