missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MITD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17264-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
MITD1 may play a role in endosomal protein transport.Specifica
MITD1 | |
Polyclonal | |
Unconjugated | |
MITD1 | |
MITD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
129531, 363219, 69028 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5I0J5, Q8VDV8, Q8WV92 | |
Mitd1 | |
MITD1 Fusion Protein Ag10998 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |