Learn More
Sartorius Confezioni pipette multipack MLINE™
Per ricaricare o impostare una nuova stazione di pipettaggio. I multipack di pipette Sartorius mLINE™ combinano in modo pratico un set di pipette meccaniche mLINE con appositi vassoi per puntali e utili accessori. Disponibili in confezioni da 3, 4 e 5 pipette con puntali associati
Marca: Sartorius LH-725664
Dettagli aggiuntivi : Peso : 1.20000kg
Descrizione
Multipacks to match every need- Affordable options to get a set of quality pipettes with useful accessories
- Ideal for renewing an existing pipette collection or setting up a new pipetting station
- Includes a linear pipette rack, calibration tool, tube opener and color-coding caps
- Collections include up to five different volumes of single channel pipettes, from 10μL to 5mL; the 3+1 pack includes a 350μL eight-channel pipette
- Offer excellent ergonomics, performance, precision and safety in manual pipetting
- Requires exceptionally light pipetting and tip ejection forces
- Specifically designed to prevent Repetitive Strain Injury (RSI) in repetitive pipetting
- Minimized risk of contamination with Safe-Cone Filters and full autoclavability
- Increased safety with volume lock, preventing accidental volume changes while pipetting
- Designed for the best possible fitting and sealing for the highest possible accuracy and precision
- Tips are RNase, DNase and endotoxin free and compliant with ISO 9001 and ISO 13485
- Environment-friendly materials and fully autoclavable to reduce waste
Specifica
Combines mLINE pipets and Optifit Tips with for safe, precise pipetting | |
Repetitive Manual Pipetting | |
mLINE | |
Repetitive manual pipetting | |
5 pipettes with 5 boxes of Optifit Trays |
Autoclavable | |
1 | |
Pipette Package | |
Pipets: 20μL, 100μL, 200μL, 1000μL and 5000μL, single channel Optifit Tip tray boxes: 3 x 200μL, 1000μL and 5000μL Linear stand and accessories |