missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLN51/CASC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18047-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. The immunoproteasome contains an alternate regulator, referred to as the 11S regulator or PA28, that replaces the 19S regulator. Three subunits of the 11S regulator have been identified. This gene encodes the gamma subunit of the 11S regulator. Six gamma subunits combine to form a homohexameric ring. Two transcript variants encoding different isoforms have been identified.Specifica
MLN51/CASC3 | |
Polyclonal | |
Unconjugated | |
CASC3 | |
BTZ, CASC3, MLN 51, MLN51, Protein barentsz, Protein CASC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192160, 22794, 259170 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15234, Q8K3W3, Q8K3X0 | |
Casc3 | |
MLN51/CASC3 Fusion Protein Ag12694 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |