Learn More
MMAA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557917
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: EMWDKMKDFQ DLMLASGELT AKRRKQQKVW MWNLIQESVL EHFRTHPTVR EQIPLLEQKV LIGALSPGLA ADFL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
The protein encoded by this gene is involved in the translocation of cobalamin into the mitochondrion, where it is used in the final steps of adenosylcobalamin synthesis. Adenosylcobalamin is a coenzyme required for the activity of methylmalonyl-CoA mutase. Defects in this gene are a cause of methylmalonic aciduria.Specifica
MMAA | |
Polyclonal | |
Unconjugated | |
MMAA | |
2810018E08Rik; AI840684; cblA; methylmalonic aciduria (cobalamin deficiency) cblA type; methylmalonic aciduria (cobalamin deficiency) type A; methylmalonic aciduria type A homolog, mitochondrial; Methylmalonic aciduria type A protein, mitochondrial; MMAA; mutant adenosylcobalamin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
166785 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVH4 | |
MMAA | |
Recombinant protein corresponding to Human MMAA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only