Learn More
MMACHC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA521798
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Raji, mouse brain. Predicted reactivity: Mouse (85%), Rat (86%), Pig (91%), Rhesus Monkey (97%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The exact function of the MMACHC protein is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Specifica
MMACHC | |
Polyclonal | |
Unconjugated | |
MMACHC | |
1810037K07Rik; cblC; Cyanocobalamin reductase (cyanide-eliminating); methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria; methylmalonic aciduria and homocystinuria type C protein; Methylmalonic aciduria and homocystinuria type C protein homolog; methylmalonic aciduria cblC type, with homocystinuria; Mmachc; RGD1310806; RP11-291L19.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25974, 67096 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9CZD0, Q9Y4U1 | |
MMACHC | |
Recombinant fragment corresponding to a region within amino acids 1 and 218 of MMACHC (Uniprot ID#Q9Y4U1) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |