Learn More
MMADHC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557991
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: TLPDVLAEPL SSERHEFVMA QYVNEFQGND APVEQEINSA ETYFESARVE CAIQTCPELL RKDFESLFPE VANGKLMILT VTQKTKNDMT VWSEEV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Involved in cobalamin metabolism.Specifica
MMADHC | |
Polyclonal | |
Unconjugated | |
MMADHC | |
2010311D03Rik; AI314967; C2orf25; CblD; CL25022; HSPC161; likely ortholog of H. sapiens chromosome 2 open reading frame 25 (C2orf25); methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria; methylmalonic aciduria and homocystinuria type D homolog, mitochondrial; Methylmalonic aciduria and homocystinuria type D protein, mitochondrial; methylmalonic aciduria and homocystinuria, cblD type; Mmadhc; My011; protein C2orf25, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27249 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H3L0 | |
MMADHC | |
Recombinant protein corresponding to Human MMADHC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only