Learn More
MMGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556905
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KDMDATSELK NKTFDTLRNH PSFYVFNHRG RVLFRPSDTA NSSNQDALSS NTSLKL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Mmgt1 encodes a protein that that mediates Mg(2+) transport.Specifica
MMGT1 | |
Polyclonal | |
Unconjugated | |
MMGT1 | |
9630048L06Rik; BC032271; EMBL:AAI68927.1}; EMC5; ER membrane protein complex subunit 5; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250; Mir5116; mir-5116; Mmgt1; mmgt1 {ECO:0000250; RGD1566339; Tmem32; transmembrane protein 32; transmembrane protein 32 {ECO:0000312; UniProtKB:Q8K273} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93380 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4V1 | |
MMGT1 | |
Recombinant protein corresponding to Human MMGT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only