missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA516566
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
PA5-16566 targets MMP-13 (Collagenase-3) in WB applications and shows reactivity with Human and Rat samples. The PA5-16566 immunogen is human recombinant MMP-13 protein.
MMP13 Polyclonal antibody specifically detects MMP13 in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifica
MMP13 | |
Polyclonal | |
Unconjugated | |
MMP13 | |
cb1034; Clg; CLG3; Collagenase; collagenase 3; collagenase 3a; matrix metalloproteinase 13a; collagenase-1; Collagenase3; Collagenase-3; collagenase-3 precursor; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase-13; matrix metalloproteinase-13; MMP-13; interstitial protein; MMP; Mmp1; MMP13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13 | |
Rabbit | |
Protein A | |
RUO | |
171052, 4322 | |
4° C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P23097, P45452 | |
MMP13 | |
Human recombinant MMP-13 protein | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |