missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528219
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: HeLa. Predicted reactivity: Mouse (91%), Rat (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene is considered a member of the membrane-type MMP (MT-MMP) subfamily. However, this protein is unique among the MT-MMP's in that it is a GPI-anchored protein rather than a transmembrane protein. The protein activates MMP-2 by cleavage.Specifica
MMP17 | |
Polyclonal | |
Unconjugated | |
MMP17 | |
matrix metallo protease; matrix metallopeptidase 17; matrix metallopeptidase 17 (membrane-inserted); matrix metalloproteinase 17; matrix metalloproteinase 17 (membrane-inserted); matrix metalloproteinase-17; membrane type-4 matrix metalloproteinase; membrane-type matrix metalloproteinase 4; Membrane-type-4 matrix metalloproteinase; MMP; Mmp17; MMP-17; MMPs; MT4MMP; MT4-MMP; MT-MMP 4; MTMMP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.74 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9ULZ9 | |
MMP17 | |
Recombinant fragment corresponding to a region within amino acids 109 and 438 of Human MMP17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |