missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA125234
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival.Specifica
MMP21 | |
Polyclonal | |
Unconjugated | |
MMP21 | |
b2b2458Clo; b2b873Clo; HTX7; matrix metallo protease; matrix metallopeptidase 21; matrix metallopeptidase 21 L homeolog; matrix metalloproteinase 21; matrix metalloproteinase-21; mmp; mmp21; MMP-21; mmp21.L; mmp21-A; MMPs; XELAEV_18034590mg; XMMP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118856, 214766, 293573, 378572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.01M PBS with 50% glycerol and 0.05% sodium azide | |
O93470, Q8K3F2, Q8N119 | |
MMP21, mmp21.L | |
Synthetic peptide corresponding to the hinge domain of human MMP-21. | |
50 μg | |
Primary | |
Human, Mouse, Rat, X. laevis | |
Antibody | |
IgG |