missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP23 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530726
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MMP23B (matrix metallopeptidase 23B) is also named as MMP21, MMP22. It is a regulator in matrix remodeling and may be playing a role in enchondral bone formation and in regulation of cranial suture closure. It has 3 isoforms produced by alternative splicing with the molecular weight of 44 kDa, 48 kDa and 46 kDa, respectively. It also can be N-glycosylated.Specifica
MMP23 | |
Polyclonal | |
Unconjugated | |
MMP23B | |
Femalysin; matrix metallo protease; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; matrix metalloproteinase-21; Matrix metalloproteinase-22; matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; MMPs; RP11-345P4.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8510, 8511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75900 | |
MMP23B | |
Recombinant fragment corresponding to a region within amino acids 92 and 362 of Human MMP23B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |