Learn More
MMP3 Monoclonal Antibody (4F10), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517123
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
MA5-17123 targets MMP3 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17123 immunogen is purified recombinant fragment of human MMP3 expressed in E. Coli. MA5-17123 detects MMP3 which has a predicted molecular weight of approximately 54kDa.
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifica
MMP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P08254, P28862 | |
MMP3 | |
Purified recombinant fragment of human MMP3 expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
4F10 | |
Unconjugated | |
MMP3 | |
CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
Mouse | |
Protein G | |
RUO | |
17392, 4314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |