missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528246
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Can degrade fibrillar type I, II, and III collagens.Specifica
MMP8 | |
Polyclonal | |
Unconjugated | |
MMP8 | |
BB138268; CLG1; Collagenase; Collagenase 2; collagenase-2; HNC; matrix metallo protease; matrix metallopeptidase 8; matrix metallopeptidase 8 (neutrophil collagenase); matrix metalloproteinase 8; matrix metalloproteinase 8 (neutrophil collagenase); Matrix metalloproteinase8; matrix metalloproteinase-8; MMP; MMP 8; Mmp8; MMP-8; MMPs; Neutrophil collagenase; neutrophil collagenease; PMN leukocyte collagenase; PMNL CL; PMNL collagenase; PMNLCL; PMNL-CL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P22894 | |
MMP8 | |
Recombinant fragment corresponding to a region within amino acids 96 and 467 of Human MMP8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |