missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNAR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA519605
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
For Western Blot, this antibody has non-specific bands at 76 kDa. This antibody is predicted to react with mouse and rat based on sequence homology.
PELP1 is a coactivator of estrogen receptor -mediated transcription and a corepressor of other nuclear hormone receptors and sequence-specific transcription factors .Specifica
MNAR | |
Polyclonal | |
Unconjugated | |
Q8IZL8 | |
PELP1 | |
Synthetic peptide conjugated to KLH derived from within residues 400 - 500 of Human MNAR. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PELP1 | |
HMX3, MNAR, P160, modulator of non-genomic activity of estrogen receptor; modulator of nongenomic activity of estrogen receptor; proline and glutamic acid rich nuclear protein; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; transcription factor HMX3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |