Learn More
MNF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558659
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: NYYKHKYPRP RDTSFSGLSL EEYKLILSTD TLEELKEIDK GMWKKLQEKF APKGPEADDK A Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 77%.
This gene encodes a nucleoid protein localized to the mitochondria inner membrane. The encoded protein affects regulation of insulin secretion, mitochondrial ATP production, and myogenesis through modulation of mitochondrial respiratory chain activity.Specifica
MNF1 | |
Polyclonal | |
Unconjugated | |
UQCC2 | |
2900010M23Rik; bA6B20.2; breast cancer-associated protein SGA-81M; C6orf125; Cbp6; cytochrome B protein synthesis 6 homolog; M19; mitochondrial nucleoid factor 1; Mitochondrial protein M19; MNF1; RGD1306917; ubiquinol-cytochrome c reductase complex assembly factor 2; ubiquinol-cytochrome-c reductase complex assembly factor 2; UQCC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84300 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRT2 | |
UQCC2 | |
Recombinant protein corresponding to Human MNF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only