Learn More
Köttermann Mobile Cabinet with One Door
This Mobile Cabinet with One Door offers versatile storage space with a single door to protect stored items.
Marca: Köttermann 315-00071
Dettagli aggiuntivi : Peso : 31.38480kg
Descrizione
- Versatile storage space
- Mobile with lockable wheels
- Features one door to protect stored items
- Many sizes and configurations available
Specifica
None | |
None | |
516 x 600 x 740 mm | |
Hinged | |
Mobile | |
Steel | |
1 | |
Steel |
Right | |
Yes | |
Plastic with Brakes | |
1 | |
Epoxy Coated | |
No | |
White |