missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Amphiregulin Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB989-500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Amphiregulin Monoclonal specifically detects Amphiregulin in Mouse samples. It is validated for Western Blot.
Specifica
| Amphiregulin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 374 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 206220 | |
| Western Blot 1 ug/mL | |
| P31955 | |
| AREG | |
| E. coli-derived recombinant mouse Amphiregulin Ser94-Lys191 Accession # P31955 | |
| 500 μg | |
| Primary | |
| Detects mouse Amphiregulin in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Amphiregulin, rhEGF, rhHB-EGF, rhNRG-1 alpha , rhNRG-1-beta 1, rhTGF-alpha , recombinant mouse (rm) BTC, rmEpigen, or rmEpiregulin. | |
| Mouse | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto