Learn More
Mouse anti-Rat and anti-Hamster Ig, κ Light Chain, Clone: RG7/7.6, Secondary Antibody, BD
Marca: BD Biosciences 550336
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The RG7/7.6 antibody reacts strongly with rat Igs bearing - light chain of the Igk-1[a]allotype (e.g., AO, AUG, BN, BUF, LEW, LOU, PVG, WAG, WF), weakly with Igk-1[b] light chains (e.g., ACI, COP, DA, F344, SHR), and it does not react with λ light chain nor heavy chains. At the time that this antibody was characterized, the Igk-1[a] allotype was designated - 1b, and Igk-1[b] was -1a; the current nomenclature was standardized at the Fourth International Workshop on Allogenic Systems in the Rat. The RG7/7.6 antibody also strongly cross-reacts with
Syrian and Armenian hamster Ig - light chain. This antibody is routinely tested by immunohistochemical analysis. Other applications were tested at BD Biosciences Pharmingen during
antibody development only or reported in the literature.
Host Species: Mouse
Clone: RG7/7.6
Isotype: IgG2a κ
Species Reactivity: Rat
Immunogen: Pooled Rat IgG
Immunohistochemistry, Immunohistochemistry (Frozen)
Specifica
Mouse anti-Rat and anti-Hamster Ig, κ Light Chain | |
RG7/7.6 | |
Mouse | |
RUO | |
Secondary | |
IgG2a κ |
Monoclonal | |
Biotin | |
1 mL | |
Immunology | |
Hamster, Rat |