Learn More
Mouse anti-Rat IgG1, Clone: RG11/39.4, FITC, Secondary Antibody, BD
Marca: BD Biosciences 553892
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The RG11/39.4 antibody reacts specifically with the Fc region of rat IgG1. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: RG11/39.4
Isotype: IgG2a, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Frozen)
Specifica
Mouse anti-Rat IgG1 | |
RG11/39.4 | |
FITC | |
0.5 mg | |
Immunology | |
Rat |
Monoclonal | |
0.5mg/mL | |
Mouse | |
RUO | |
Secondary | |
IgG2a κ |