Learn More
ACROBiosystems Mouse CEACAM-5 Protein, Fc Tag
Marca: ACROBiosystems CE5-M5253-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) is also Carcinoembryonic antigen (CEA), Meconium antigen 100, CD antigen CD66e, CEACAM5 belongs to the immunoglobulin superfamily and CEA family. CEACAM5 contains seven Ig-like (immunoglobulin-like) domains. CEACAM5 is homodimer protein which binding of E.coli Dr adhesins leads to dissociation of the homodimer. CEACAM5 is cell surface glycoprotein that plays a role in cell adhesion and in intracellular signaling. CEACAM5 is receptor for E.coli Dr adhesins.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 129.1 kDa.Specifica
129.1 kDa | |
Mouse CEACAM-5, Fc Tag (CE5-M5253) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Glu 947 (Accession # Q3UKK2-1). | |
-20°C | |
CEACAM-5 | |
Mouse | |
HEK293 | |
Bio-Markers & CD Antigens |
1 mg | |
Gln 35 - Glu 947 | |
CEACAM-5,CD66e,CEA,Meconium antigen 100 | |
Unconjugated | |
Fc Tag | |
Powder |