missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse GUCY2C / Guanylyl cyclase C Protein, His Tag
Marca: ACROBiosystems GUC-M52H3-100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GUCY2C (Guanylyl Cyclase C), also known as heat-stable enterotoxin receptor, is a type I transmembrane protein of the guanylate cyclase (gc) family that signal by producing cGMP. Guanylate cyclase C (GUCY2C) and its hormones guanylin and uroguanylin have recently emerged as one paracrine axis defending intestinal mucosal integrity against mutational, chemical, and inflammatory injury. GUCY2C murine CAR-T cells recognized and killed human colorectal cancer cells endogenously expressing GUCY2C. Thus, we have identified a human GUCY2C-specific CAR-T cell therapy approach that may be developed for the treatment of GUCY2C-expressing metastatic colorectal cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 60-100 kD under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
49.0 kDa | |
Mouse GUCY2C, His Tag (GUC-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Met 433 (Accession # Q3UWA6-1). | |
-20°C | |
GUCY2C | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Val 20 - Met 433 | |
GUCY2C,GUC2C,STAR,STA receptor,hSTAR,GC-C | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |