Learn More
Mouse IgG VisUCyte HRP Polymer Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems VC001-050
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Immunohistochemistry.
Specifica
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse IgG | |
| 50 mL (2 x 25 mL) | |
| Secondary | |
| Detects heavy ( gamma ) chains on mouse IgG and light chains on all mouse immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin mouse serum proteins or serum proteins from bovine, horse, human, pig or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC. | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
| Immunohistochemistry | |
| HRP Polymer | |
| Supplied as a solution in PBS with preservative. | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Mouse | |
| IgG |