Learn More
ACROBiosystems Mouse Mesothelin / MSLN (298-600) Protein, His Tag
MALS verified
Marca:  ACROBiosystems MSN-M52H3-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106511
  • 3357.52€ / 1mg
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 42-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_061345.1
1 mg
Asp 298 - Ser 600
MSLN,Mesothelin,MPF
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
36.0 kDa
Mouse Mesothelin (298-600), His Tag (MSN-M52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).
-20°C
Mesothelin
Mouse
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens