Learn More
ACROBiosystems Mouse Mesothelin / MSLN (298-600) Protein, His Tag
MALS verified
Marca: ACROBiosystems MSN-M52H3-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 42-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_061345.1 | |
100 μg | |
Asp 298 - Ser 600 | |
MSLN,Mesothelin,MPF | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
36.0 kDa | |
Mouse Mesothelin (298-600), His Tag (MSN-M52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1). | |
-20°C | |
Mesothelin | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |