Learn More
MRP1 / ABCC1 Mouse, Clone: MRP1/1343, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 4363-MSM1-P0
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| MRP1 / ABCC1 | |
| Cytoplasmic and Cell Surface | |
| MRP1/1343 | |
| Unconjugated | |
| XP_016878731 | |
| Mouse | |
| 190kDa | |
| Protein A/G Purified | |
| RUO | |
| 4363 | |
| IgG1 |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| ABCC1 | |
| Recombinant human full-length ABCC1 | |
| A549 cells. Colon, Endometrium, Ovary. Testis or Tonsil. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |