missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17280-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3).Specifica
MRPS27 | |
Polyclonal | |
Unconjugated | |
MRPS27 | |
KIAA0264, MRP S27, MRPS27, S27mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
218506, 23107, 361883 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BK72, Q92552 | |
Mrps27 | |
MRPS27 Fusion Protein Ag11047 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |