missing translation for 'onlineSavingsMsg'
Learn More

MUC16/CA125 Mouse anti-Human, Clone: 2B11B10, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60261-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897305

  • 325.38€ / 150µL

Spedizione stimata: 04-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The antibody recognizes the C-term of MUC16.

CA 125 determit is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
Specifica

Specifica

MUC16/CA125
Monoclonal
0.7 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WXI7
MUC16
Peptide
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
2B11B10
Unconjugated
MUC16
CA 125, CA125, MUC 16, MUC16, Mucin 16
Mouse
Protein G
RUO
94025
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali