missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 55489-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is specific to MUC8.
Mucin glycoproteins (Mucins) are major constituents of the glycocalyx that covers mucosal epithelium. MUC8 is one of the major mucins in the ethmoid mucosa and is upregulated by chronic inflammation. TNFa, IL-1b and a combination of both can significantly increase Mucin 8 mRNA levels, suggesting that a mixture of inflammatory mediators can synergistically increase secretion of mucin in human nasal epithelium.Specifica
MUC8 | |
Polyclonal | |
Unconjugated | |
MUC8 | |
MUC8 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MUC8, mucin 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100129528 | |
-20°C | |
Liquid |