Learn More
Honeywell™ Mutex E Light Protective Clothing
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
Pezzo
Descrizione
- Antistatic
- Silicone and latex free
- Excellent ergonomics and fitting
- 3-piece hood
- Special crotch design
- Outside flap and dual slider sipper
- Breathability
- 3-layer polypropylene
Specifica
Specifica
| Product Type | Chemical-resistant Coveralls |
| Size | 2X-Large |
| Sesso | Unisex |
| Materiale | 3-layer Polypropylene |
| Colore | White |
| Classe di protezione | Protection type: 5, 6 |
| Certifications/Compliance (Europe) | Ec Category PPE: 3, EC Certificate: 78815277, EC Attestation: 78815277 |
| Spessore | 56 gsm |