missing translation for 'onlineSavingsMsg'
Learn More

MUTYH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19650-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899124

  • 140.00€ / 20µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.
Specifica

Specifica

MUTYH
Polyclonal
Unconjugated
MUTYH
hMYH, MutY homolog, mutY homolog (E. coli), MUTYH, MYH
Rabbit
Antigen Affinity Chromatography
RUO
170841, 4595, 70603
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R5G2, Q99P21, Q9UIF7
MUTYH
MUTYH Fusion Protein Ag7526
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificates
Special Offers

Special Offers