missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10731-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody can detect both of isoforms around 57 kDa and 35 kDa.
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifica
| MYOT | |
| Polyclonal | |
| Unconjugated | |
| MYOT | |
| 57 kDa cytoskeletal protein, LGMD1, LGMD1A, MYOT, myotilin, TTID | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291605, 58916, 9499 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9JIF9, Q9UBF9 | |
| Myot | |
| MYOT Fusion Protein Ag1112 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto