Learn More
MYOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifica
Antigene MYOT
Concentrazione 0.18 mg/mL
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
16877303
Vedere i documenti
Proteintech
10731-1-AP-20UL
20 μL
145.00€
20µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
16867303
Vedere i documenti
Proteintech
10731-1-AP-150UL
150 μL
410.00€
150µL
Visualizzare prodotti alternativi
Risparmia fino a 
»
Descrizione
Descrizione
This antibody can detect both of isoforms around 57 kDa and 35 kDa.
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifica
Specifica
MYOT
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
Q9JIF9, Q9UBF9
291605, 58916, 9499
MYOT Fusion Protein Ag1112
Primary
-20°C
MYOT
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
57 kDa cytoskeletal protein, LGMD1, LGMD1A, MYOT, myotilin, TTID
Myot
IgG
Antigen Affinity Chromatography