missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dichloro-3-pyridyl)thiourea, 98+%, Thermo Scientific™

Catalog Number 15455947
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1421261-48-4
C6H5Cl2N3S
222.087
MFCD22682806
BUCIELRKJVZURG-UHFFFAOYSA-N
n-2,6-dichloro-3-pyridyl thiourea
73995147
(2,6-dichloropyridin-3-yl)thiourea
C1=CC(=NC(=C1NC(=S)N)Cl)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1421261-48-4
C6H5Cl2N3S
222.087
MFCD22682806
BUCIELRKJVZURG-UHFFFAOYSA-N
n-2,6-dichloro-3-pyridyl thiourea
73995147
(2,6-dichloropyridin-3-yl)thiourea
C1=CC(=NC(=C1NC(=S)N)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-(2,6-Dichloro-3-pyridyl)thiourea
Punto di fusione 220°C to 224°C
Quantità 250 mg
Peso formulazione 222.09
Percent Purity ≥98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'