missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecanoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-203442
Click to view available options
Quantità:
10 g
25 g
50 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
638-53-9
C13H26O2
214.35
MFCD00002741
SZHOJFHSIKHZHA-UHFFFAOYSA-N
n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16
12530
CHEBI:45919
tridecanoic acid
CCCCCCCCCCCCC(=O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
638-53-9
C13H26O2
214.35
MFCD00002741
SZHOJFHSIKHZHA-UHFFFAOYSA-N
n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16
12530
CHEBI:45919
tridecanoic acid
CCCCCCCCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico n-Tridecanoic acid
Nota nome 97%
CAS min. % 96.0
CAS max % 100.0
Punto di fusione 40°C to 45°C
Colore White to Yellow
Punti di ebollizione 236°C (100.0 mmHg)
Punto d'infiammabilità 113°C
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97%
Imballaggio Glass bottle
Formula lineare CH3(CH2)11CO2H
Quantità 10 g
Beilstein 02, 364
Informazioni di solubilità Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Peso formulazione 214.35
Percent Purity 97%
Forma fisica Crystalline Chunks, Powder or Flakes
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'