missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19601-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Transcriptional control is in part regulated by interactions between DNA-bound transcription factors, such as Egr-1/NGFI-A, and coregulatory proteins, such as NAB (for NGFI-A-binding proteins). The evolutionarily conserved NAB proteins, NAB1 and NAB2, are corepressors of EGF-1/NGFI-A. Both NAB1 and NAB2 contain an amino terminal NAB conserved domain 1 (NCB1), which is required for binding NGFI-A, and a carboxy terminal NCD2 domain, which is responsible for the repressor function of NAB proteins. NAB2 is principally localized in the nucleus and may play a role in the downregulation of NGFI-A activity as well as in controlling fundamental processes such as cell division, differentiation and apoptosis. NAB2 has a predicted molecular mass of 56 kDa and localizes to chromosome 12q13.3-14.1, a region that is rearranged in several solid tumors, lipomas and liposarcomas.Specifica
NAB2 | |
Polyclonal | |
Unconjugated | |
NAB2 | |
EGR 1 binding protein 2, MADER, NAB2, NGFI A binding protein 2, Protein MADER | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17937, 314910, 4665 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15742, Q61127 | |
Nab2 | |
NAB2 Fusion Protein Ag6346 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |