Learn More
Cytiva (formerly Pall Lab) NAB Nanosep™ Centrifugal Devices for Nucleic Acid Isolation
The Nucleic Acid Binding Nanosep spin device is designed for the specific purpose of extracting, isolating and purifying genomic DNA (gDNA), RNA and plasmid DNA (pDNA) from various starting materials such as bacteria, yeast, mammalian cultured cells, and plants, for molecular research applications.
Marca: Cytiva (formerly Pall Lab) ODNABC34
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
Don't let left-over reagents go to waste, use them with the Nucleic Acid Binding™ universal spin device. This centrifugal device can also be used with buffers prepared inhouse or off-the-shelf. A variety of protocols are available to download for different starting materials.
Downstream Applications:
Purified Plasmid DNA
- Restriction digest
- Cloning
- Sanger sequencing
Purified Genomic DNA
- qPCR
- Next Generation Sequencing
Purified Total RNA
- RT-PCR
- cDNA library construction
This product is not approved for use in medical, clinical, surgical or other patient applications.
Specifica
EFA: 0.3 cm2 | |
Non-sterile | |
100/Pk. |
Glass Fiber | |
Polypropylene | |
50 to 500 μL |