Learn More
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
Marca: Thermo Scientific™ C4010-S92
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
- Two-compartment benchtop storage tray
- I-D™ vials feature write-on patch with graduations
- All items remain handy and dust-free
- Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifica
Light Blue | |
Clear | |
Yes | |
PTFE/Silicone, Pre-slit | |
No |
Clear Glass; Silanized; Light Blue Cap, Pre-Slit Blue PTFE/White SiliconeSeptum; 2mL | |
PTFE, Silicone | |
2 mL | |
Silanized |