missing translation for 'onlineSavingsMsg'
Learn More

NDOR1/NR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11404-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898453

  • 325.38€ / 150µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants.
Specifica

Specifica

NDOR1/NR1
Polyclonal
Unconjugated
NDOR1
NDOR1, Novel reductase 1, NR1
Rabbit
Antigen Affinity Chromatography
RUO
27158, 311799, 78797
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2AI05, Q9UHB4
NDOR1
NDOR1/NR1 Fusion Protein Ag1909
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali